Learn More
Medchemexpress LLC HY-15427 5mg Medchemexpress, GDC-0834 CAS:1133432-49-1 Purity:>98%
SDP
Catalog No. 502030624
Encompass_Preferred
Medchemexpress, HY-15427 5mg GDC-0834 CAS:1133432-49-1 GDC-0834 is a potent and selective BTK inhibitor. GDC-0834 inhibits BTK with an in vitro IC50 of 5.9 and 6.4 nM in biochemical and cellular assays, respectively, and in vivo IC50 of 1.1 and 5.6 μM in mouse and rat, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0624 Supplier Medchemexpress LLC Supplier No. HY154275MG
Medchemexpress, HY-15427 5mg GDC-0834 CAS:1133432-49-1 GDC-0834 is a potent and selective BTK inhibitor. GDC-0834 inhibits BTK with an in vitro IC50 of 5.9 and 6.4 nM in biochemical and cellular assays, respectively, and in vivo IC50 of 1.1 and 5.6 μM in mouse and rat, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.