Learn More
Medchemexpress LLC HY-50864 5mg , GDC-0879 CAS:905281-76-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY508645MG
Encompass_Preferred
HY-50864 5mg GDC-0879 CAS: 905281-76-7 GDC-0879 is a potent and selective B-Raf inhibitor with an IC50 of 0.13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1081