Learn More
Selleck Chemical LLC GDC-0879 5mg 905281-76-7 AR-00341677
SDP
Catalog No. 508426
Encompass_Preferred
GDC-0879 (AR-00341677) is a novel, potent, and selective B-Raf inhibitor with IC50 of 0.13 nM in A375 and Colo205 cells with activity against c-Raf as well; no inhibition known to other protein kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-842-6 Supplier Selleck Chemical LLC Supplier No. S11045MG
GDC-0879 (AR-00341677) is a novel, potent, and selective B-Raf inhibitor with IC50 of 0.13 nM in A375 and Colo205 cells with activity against c-Raf as well; no inhibition known to other protein kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.