Learn More
Apexbio Technology LLC GDC-0941 957054-30-7 200mg
SDP
Catalog No. 5000062499
Encompass_Preferred
GDC-0941 (CAS 957054-30-7) is a selective small-molecule inhibitor of class I phosphoinositide 3-kinase (PI3K) a critical component involved in the PI3K/Akt signaling pathway frequently dysregulated in cancer By competitively binding to the ATP-binding site of PI3K GDC-0941 inhibits generation of the downstream signaling molecule phosphatidylinositol-3 4 5-trisphosphate (PIP3) Exhibiting high selectivity against p110 / (IC50 approx 3 nM) and moderate selectivity for p110 (33 nM) and p110 (75 nM) GDC-0941 has demonstrated antiproliferative activity across diverse tumor cell lines in vitro (e g A2780 PC3 U87MG) and tumor growth inhibition in xenograft cancer models supporting its utility in cancer research
Catalog No. 50-000-62499 Supplier Apexbio Technology LLC Supplier No. A8210200
GDC-0941 (CAS 957054-30-7) is a selective small-molecule inhibitor of class I phosphoinositide 3-kinase (PI3K) a critical component involved in the PI3K/Akt signaling pathway frequently dysregulated in cancer By competitively binding to the ATP-binding site of PI3K GDC-0941 inhibits generation of the downstream signaling molecule phosphatidylinositol-3 4 5-trisphosphate (PIP3) Exhibiting high selectivity against p110 / (IC50 approx 3 nM) and moderate selectivity for p110 (33 nM) and p110 (75 nM) GDC-0941 has demonstrated antiproliferative activity across diverse tumor cell lines in vitro (e g A2780 PC3 U87MG) and tumor growth inhibition in xenograft cancer models supporting its utility in cancer research