Learn More
Selleck Chemical LLC Pictilisib (GDC-0941) 5mg 957054-30-7 RG7321
SDP
Catalog No. 508516
Encompass_Preferred
Pictilisib (GDC-0941, RG7321) is a potent inhibitor of PI3K?/? with IC50 of 3 nM in cell-free assays, with modest selectivity against p110? (11-fold) and p110? (25-fold). Pictilisib (GDC-0941) induces autophagy and apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-851-6 Supplier Selleck Chemical LLC Supplier No. S10655MG
Add to Cart
Add to Cart
Pictilisib (GDC-0941, RG7321) is a potent inhibitor of PI3K?/? with IC50 of 3 nM in cell-free assays, with modest selectivity against p110? (11-fold) and p110? (25-fold). Pictilisib (GDC-0941) induces autophagy and apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.