Learn More
Selleck Chemical LLC Cobimetinib (GDC-0973) 5mg 934660-93-2 RG7420
SDP
Supplier:  Selleck Chemical LLC S80415MG
Encompass_Preferred
Cobimetinib (GDC-0973, RG7420) is a potent and highly selective MEK1 inhibitor with IC50 of 4.2 nM, showing more than 100-fold selectively for MEK1 over MEK2 and showed no significant inhibition when tested against a panel of more than 100 of serine-threonine and tyrosine kinases. Cobimetinib induces apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1033334