Learn More
Apexbio Technology LLC GDC-0994 1453848-26-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062761
Encompass_Preferred
GDC-0994 (CAS 1453848-26-4) is a selective small-molecule inhibitor targeting extracellular signal-regulated kinases 1 and 2 (ERK1/2) components of the MAP kinase signaling cascade downstream of RAS/RAF/MEK ERK1/2 mediates cell proliferation and differentiation frequently activated due to oncogenic mutations in RAS and BRAF GDC-0994 shows inhibition of ERK1 and ERK2 with IC50 values of 1 1 and 0 3 nM respectively In preclinical models bearing KRAS or BRAF mutant tumor xenografts oral administration of GDC-0994 suppresses ERK phosphorylation and downstream signaling demonstrating significant anti-tumor activity GDC-0994 is a potent tool for studying ERK signaling in cancer research
Catalog No. 50-000-62761 Supplier Apexbio Technology LLC Supplier No. B58175.1
GDC-0994 (CAS 1453848-26-4) is a selective small-molecule inhibitor targeting extracellular signal-regulated kinases 1 and 2 (ERK1/2) components of the MAP kinase signaling cascade downstream of RAS/RAF/MEK ERK1/2 mediates cell proliferation and differentiation frequently activated due to oncogenic mutations in RAS and BRAF GDC-0994 shows inhibition of ERK1 and ERK2 with IC50 values of 1 1 and 0 3 nM respectively In preclinical models bearing KRAS or BRAF mutant tumor xenografts oral administration of GDC-0994 suppresses ERK phosphorylation and downstream signaling demonstrating significant anti-tumor activity GDC-0994 is a potent tool for studying ERK signaling in cancer research