Learn More
Selleck Chemical LLC Ravoxertinib (GDC-0994) 5mg 1453848-26-4
SDP
Supplier:  Selleck Chemical LLC S75545MG
Encompass_Preferred
Ravoxertinib (GDC-0994) is a potent, orally available and highly selective ERK1/2 inhibitor with IC50 of 1.1 nM and 0.3 nM, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1185172