Learn More
Medchemexpress LLC HY-15947 50mg , GDC-0994 Ravoxertinib CAS:1453848-26-4 Purity:98%
SDP
Catalog No. 501493653
Encompass_Preferred
Medchemexpress, HY-15947 50mg GDC-0994 Ravoxertinib CAS:1453848-26-4 Formula:C21H18ClFN6O2 IC50: 6.1 nM (ERK1), 3.1 nM (ERK2), 12 nM (p90RSK) Purity:98% GDC-0994 is an orally bioavailable inhibitor selective for ERK kinase activity with IC 50 of 6.1 nM and 3.1 nM for ERK1 and ERK2 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3653 Supplier Medchemexpress LLC Supplier No. HY1594750MG
Medchemexpress, HY-15947 50mg GDC-0994 Ravoxertinib CAS:1453848-26-4 Formula:C21H18ClFN6O2 IC50: 6.1 nM (ERK1), 3.1 nM (ERK2), 12 nM (p90RSK) Purity:98% GDC-0994 is an orally bioavailable inhibitor selective for ERK kinase activity with IC 50 of 6.1 nM and 3.1 nM for ERK1 and ERK2 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.