Learn More
TARGETMOL CHEMICALS INC GDC0084 5MG
SDP
Catalog No. 502576882
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GDC0084 (RG7666) is a PI3K inhibitor with potential antineoplastic activity. GDC0084 (RG7666) specifically inhibits PI3K in the PI3K/AKT kinase (or protein kinase B) signaling pathway thereby inhibiting the activation of the PI3K signaling pathway. This may result in the inhibition of both cell growth and survival in susceptible tumor cell populations. purity: 99%
Catalog No. 50-257-6882 Supplier TARGETMOL CHEMICALS INC Supplier No. T37055MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GDC0084 (RG7666) is a PI3K inhibitor with potential antineoplastic activity. GDC0084 (RG7666) specifically inhibits PI3K in the PI3K/AKT kinase (or protein kinase B) signaling pathway thereby inhibiting the activation of the PI3K signaling pathway. This may result in the inhibition of both cell growth and survival in susceptible tumor cell populations. purity: 99%