Learn More
Aviva Systems Biology Corp GDE1 Antibody - N-terminal region (ARP47287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDE1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GDE1. Alternate names: 363E6.2, GDE1, MIR16ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59509