Learn More
Enzo Life Sciences Growth and Differentiation factor-11 (human), (recombinant) (20µg)
SDP
Catalog No. 502014037
Encompass_Preferred
Alternative name: GDF-11, Bone morphogenetic protein 11, BMP-11. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 109/218 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4037 Supplier Enzo Life Sciences Supplier No. ENZPRT2360020
Add to Cart
Add to Cart
Alternative name: GDF-11, Bone morphogenetic protein 11, BMP-11. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 109/218 amino acids. Long Term Storage: -80°C.