Learn More
Eurofins Discoverx Recombinant Human GDF-2 (BMP-9), 10 µg. (92-1249)
Supplier:  Eurofins Discoverx 921249
Encompass
Growth differentiation factor 2 (GDF-2, BMP-9) is a member of the bone morphogenetic protein (BMP) family that plays a role in cholinergic CNS neuron differentiation and response to the neurotransmitter acetylcholine, as well as the regulation of embryonic and adult tissue cell growth and differentiation. Potent circulatory inhibitor of angiogenesis, signaling through the type I activin receptor kinase ACVRL1. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9770