Learn More
Aviva Systems Biology Corp GDF15 Antibody - N-terminal region (ARP42200_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42200_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDF15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GDF15. Alternate names: GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFBApplicationWB. Species reactivity: Human
Catalog No. 50-014-58138