Learn More
Cell Signaling Technology GDF15/MIC1 (E5Q8Q) Rabbit Monoclonal Antibody, Human Reactivity, Unconjugated, 100 μL
SDP
Catalog No. 502731139
Encompass
GDF15/MIC1 (E5Q8Q) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total GDF15/MIC1 protein. This antibody is specific to monomeric and dimeric forms of the pro-GDF15/MIC1 protein. It is produced by immunizing animals with recombinant protein specific to full-length human GDF15/MIC1 protein. The antibody is suitable for applications including Western blotting and immunohistochemistry (paraffin).
  • Validated for Western blotting (1:1000 dilution) and immunohistochemistry (paraffin) (1:50 to 1:200 dilution).
  • Specific to human species reactivity.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at −20°C.
Catalog No. 50-273-1139 Supplier Cell Signaling Technology Supplier No. 79996S
GDF15/MIC1 (E5Q8Q) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total GDF15/MIC1 protein. This antibody is specific to monomeric and dimeric forms of the pro-GDF15/MIC1 protein. It is produced by immunizing animals with recombinant protein specific to full-length human GDF15/MIC1 protein. The antibody is suitable for applications including Western blotting and immunohistochemistry (paraffin).
  • Validated for Western blotting (1:1000 dilution) and immunohistochemistry (paraffin) (1:50 to 1:200 dilution).
  • Specific to human species reactivity.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at −20°C.