Learn More
Aviva Systems Biology Corp GDI2 Antibody - C-terminal region (AVARP13037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDI2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GDI2. Alternate names: RABGDIBApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Sheep
Catalog No. 50-014-53782