Learn More
Anatrace GDN 25G
Supplier:  Anatrace GDN101 25 GM
Encompass
GDN101 Synthetic drop-in substitute for Digitonin • glyco-diosgenin, ≥ 97% (by HPLC analysis) CAS#1402423-29-3 C56H92O25 MW:1165.31
Catalog No. NC2004999
Only null left
Add to Cart
Request a Quote