Learn More
Enzo Life Sciences gDNA removal kit (250 Reactions)
SDP
Catalog No. 502013831
Encompass_Preferred
gDNA removal kit uses a heat labile dsDNase to remove gDNA from RNA preps prior to reverse transcription. The kit is based on a recombinant heat labile dsDNase which is irreversibly inactivated at low temperatures. This enables an inactivation step including heating to moderate temperatures only (58°C), which is gentle enough to preserve both quality and quantity of all present RNA. Reverse transcription can be performed in the same tube, minimizing pipetting steps and reducing hands-on time.
Catalog No. 50-201-3831 Supplier Enzo Life Sciences Supplier No. ENZKIT1360250
gDNA removal kit uses a heat labile dsDNase to remove gDNA from RNA preps prior to reverse transcription. The kit is based on a recombinant heat labile dsDNase which is irreversibly inactivated at low temperatures. This enables an inactivation step including heating to moderate temperatures only (58°C), which is gentle enough to preserve both quality and quantity of all present RNA. Reverse transcription can be performed in the same tube, minimizing pipetting steps and reducing hands-on time.