Learn More
Enzo Life Sciences GDNF (human), (recombinant) (10µg)
SDP
Catalog No. 502013936
Encompass_Preferred
Alternative name: Glial cell-derived neurotrophic factor, Astrocyte-derived trophic factor, ATF. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10 mM sodium citrate, containing 150mM NaCl. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3936 Supplier Enzo Life Sciences Supplier No. ENZPRT1450010
Alternative name: Glial cell-derived neurotrophic factor, Astrocyte-derived trophic factor, ATF. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10 mM sodium citrate, containing 150mM NaCl. Source: Produced in E.coli. Long Term Storage: -20°C.