Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDNF Human 10ug
SDP

Catalog No. 501970578
Encompass_Preferred

Recombinant human glial cell line-derived neurotrophic factor (GDNF) supplied lyophilized for research use. The product corresponds to the mature human sequence (S78-I211), is expressed in E. coli, and is provided as a tag-free preparation with high purity suitable for neuronal survival, differentiation, and signaling studies.

  • Recombinant human sequence corresponding to mature GDNF.
  • Tag free preparation to preserve native activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized format for improved stability and storage.
  • Small pack sizes suitable for assay optimization and pilot studies.
  • Stored and handled to minimize freeze-thaw and maintain activity.

Catalog No. 50-197-0578 Supplier Medchemexpress LLC Supplier No. HYP718210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human glial cell line-derived neurotrophic factor (GDNF) supplied lyophilized for research use. The product corresponds to the mature human sequence (S78-I211), is expressed in E. coli, and is provided as a tag-free preparation with high purity suitable for neuronal survival, differentiation, and signaling studies.

  • Recombinant human sequence corresponding to mature GDNF.
  • Tag free preparation to preserve native activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized format for improved stability and storage.
  • Small pack sizes suitable for assay optimization and pilot studies.
  • Stored and handled to minimize freeze-thaw and maintain activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.