Learn More
Medchemexpress LLC GDNF Human HEK293 100ug
SDP
Catalog No. 5000180443
Encompass_Preferred
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Human (HEK293) is produced in HEK293 cells and consists of 103 amino acids (R83-I185)
Catalog No. 50-001-80443 Supplier Medchemexpress LLC Supplier No. HYP73074100UG
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Human (HEK293) is produced in HEK293 cells and consists of 103 amino acids (R83-I185)