Learn More
Medchemexpress LLC GDNF Human HEK293 50ug
Catalog No. 5000181294
Encompass_Preferred
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Human (HEK293) is produced in HEK293 cells and consists of 103 amino acids (R83-I185)
Catalog No. 50-001-81294 Supplier Medchemexpress LLC Supplier No. HYP7307450UG
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Human (HEK293) is produced in HEK293 cells and consists of 103 amino acids (R83-I185)