Learn More
Medchemexpress LLC GDNF Mouse CHO 10ug
SDP
Catalog No. 5000180805
Encompass_Preferred
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Mouse (CHO) is produced in CHO cells and consists of 134 amino acids (S78-I211)
Catalog No. 50-001-80805 Supplier Medchemexpress LLC Supplier No. HYP735910UG
Glial-cell-line-derived neurotrophic factor (GDNF) a neurotrophic factor belongs to the GDNF family ligands (GFLs) promotes survival of dopamine neurons GDNF demonstrates a variety of neuroprotective roles for mammalian neurons[1] GDNF Protein Mouse (CHO) is produced in CHO cells and consists of 134 amino acids (S78-I211)