Learn More
American Radiolabeled Chemicals Inc GDP-5-8-3H A 250UC
SDP
Catalog No. 507532677
Encompass
Guanosine 5'-5',8-3H diphosphate trisodium salt (GDP) 250uCi, 25-50 Ci/mmol, sterile water, 1 mCi/ml, M.W. 509.2, Shipped in Dry Ice
Catalog No. 50-753-2677 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1736A250UC
Guanosine 5'-5',8-3H diphosphate trisodium salt (GDP) 250uCi, 25-50 Ci/mmol, sterile water, 1 mCi/ml, M.W. 509.2, Shipped in Dry Ice