Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDP-D-mannose disodium | 103301-73-1 | 10 MG
SDP

Catalog No. 5000417307
Encompass_Preferred

GDP-D-mannose disodium is an endogenous metabolite consisting of GDP-α-D-mannose and GDP-β-D-mannose. GDP-α-D-mannose acts as the donor substrate for mannosyltransferases and serves as the precursor for GDP-β-L-fucose. It exhibits competitive inhibition with respect to GTP and uncompetitive inhibition with respect to mannose-1-P. This product is intended for research use only.

  • Comprises GDP-α-D-mannose and GDP-β-D-mannose
  • Donor substrate for mannosyltransferases
  • Precursor for GDP-β-L-fucose
  • Exhibits competitive inhibition with GTP (Ki 14.7 μM)
  • Exhibits uncompetitive inhibition with mannose-1-P (Ki 115 μM)
  • Molecular weight: 649.30
  • Chemical formula: C16H23N5Na2O16P2
  • Appearance: Solid, white to off-white
  • Classified as an endogenous metabolite

Catalog No. 50-004-17307 Supplier Medchemexpress LLC Supplier No. HYN7389A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GDP-D-mannose disodium is an endogenous metabolite consisting of GDP-α-D-mannose and GDP-β-D-mannose. GDP-α-D-mannose acts as the donor substrate for mannosyltransferases and serves as the precursor for GDP-β-L-fucose. It exhibits competitive inhibition with respect to GTP and uncompetitive inhibition with respect to mannose-1-P. This product is intended for research use only.

  • Comprises GDP-α-D-mannose and GDP-β-D-mannose
  • Donor substrate for mannosyltransferases
  • Precursor for GDP-β-L-fucose
  • Exhibits competitive inhibition with GTP (Ki 14.7 μM)
  • Exhibits uncompetitive inhibition with mannose-1-P (Ki 115 μM)
  • Molecular weight: 649.30
  • Chemical formula: C16H23N5Na2O16P2
  • Appearance: Solid, white to off-white
  • Classified as an endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.