Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDP-D-mannose disodium | 103301-73-1 | 5 MG
SDP

Catalog No. 5000417438
Encompass_Preferred

GDP-D-mannose disodium is a compound containing GDP-α-D-mannose and GDP-β-D-mannose. GDP-α-D-mannose acts as a donor substrate for mannosyltransferases and is a precursor of GDP-β-L-fucose. It exhibits competitive inhibition with GTP (Ki 14.7 μM) and uncompetitive inhibition with mannose-1-P (Ki 115 μM). This product is intended for research use only.

  • Donor substrate for mannosyltransferases
  • Precursor of GDP-β-L-fucose
  • Competitive inhibition with GTP (Ki 14.7 μM)
  • Uncompetitive inhibition with mannose-1-P (Ki 115 μM)
  • White to off-white solid appearance
  • Molecular weight of 649.30
  • Molecular formula C16H23N5Na2O16P2
  • Purity of 99.68%
  • Storage at -20°C, sealed and away from moisture

Catalog No. 50-004-17438 Supplier Medchemexpress LLC Supplier No. HYN7389A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GDP-D-mannose disodium is a compound containing GDP-α-D-mannose and GDP-β-D-mannose. GDP-α-D-mannose acts as a donor substrate for mannosyltransferases and is a precursor of GDP-β-L-fucose. It exhibits competitive inhibition with GTP (Ki 14.7 μM) and uncompetitive inhibition with mannose-1-P (Ki 115 μM). This product is intended for research use only.

  • Donor substrate for mannosyltransferases
  • Precursor of GDP-β-L-fucose
  • Competitive inhibition with GTP (Ki 14.7 μM)
  • Uncompetitive inhibition with mannose-1-P (Ki 115 μM)
  • White to off-white solid appearance
  • Molecular weight of 649.30
  • Molecular formula C16H23N5Na2O16P2
  • Purity of 99.68%
  • Storage at -20°C, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.