Learn More
TARGETMOL CHEMICALS INC GDP-L-fucose 1MG
SDP
Catalog No. 502876829
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GDP-L-fucose is a nucleotide sugar widely found in fungi and plants, playing a role in both the slave and secondary remedial metabolic pathways. Purity 98.06%
Catalog No. 50-287-6829 Supplier TARGETMOL CHEMICALS INC Supplier No. T740411MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GDP-L-fucose is a nucleotide sugar widely found in fungi and plants, playing a role in both the slave and secondary remedial metabolic pathways. Purity 98.06%