Learn More
Medchemexpress LLC GDP-L-fucose disodium | 148296-47-3 | MFCD00799980 | 98.8% | 633.31 g/mol | C16H23N5Na2O15P2 | 5 MG
SDP
Catalog No. 5000223381
Encompass_Preferred
GDP-L-fucose disodium is a nucleotide sugar supplied as the disodium salt that provides the fucose moiety for biosynthesis of fucose-containing oligosaccharides. It is intended for use as a substrate in enzymatic and biochemical assays studying fucosylation and glycosylation pathways. The material is high-purity and should be stored refrigerated to preserve stability.
  • High purity suitable for biochemical assays.
  • Provides fucose moiety for enzymatic fucosylation reactions.
  • Supplied as the disodium salt for improved aqueous solubility.
  • Stable when stored at 4°C, protected from moisture and light.
  • Available in small research-scale quantities for assay development.
Catalog No. 50-002-23381 Supplier Medchemexpress LLC Supplier No. HY134433A5MG
GDP-L-fucose disodium is a nucleotide sugar supplied as the disodium salt that provides the fucose moiety for biosynthesis of fucose-containing oligosaccharides. It is intended for use as a substrate in enzymatic and biochemical assays studying fucosylation and glycosylation pathways. The material is high-purity and should be stored refrigerated to preserve stability.
  • High purity suitable for biochemical assays.
  • Provides fucose moiety for enzymatic fucosylation reactions.
  • Supplied as the disodium salt for improved aqueous solubility.
  • Stable when stored at 4°C, protected from moisture and light.
  • Available in small research-scale quantities for assay development.