Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDP-L-fucose disodium | 15839-70-0 | 99.8% | C16H25N5O15P2 | 10MG
SDP

Catalog No. 5000259287
Encompass_Preferred

GDP-L-fucose disodium is a nucleotide sugar that serves as the fucose donor in the biosynthesis of fucose-containing oligosaccharides and is used as a substrate in enzymatic and biochemical assays. The material is supplied as a high-purity solid suitable for analytical standards and enzyme reaction studies.

  • High purity (99.8%) for reliable experimental results.
  • Chemical formula C16H25N5O15P2 for precise molecular identification.
  • Supplied as a disodium salt for good water solubility and ease of use.
  • Suitable as a substrate for fucosyltransferase and glycosylation assays.
  • Available in small-scale quantities appropriate for research applications.
  • CAS number 15839-70-0 provided for unambiguous identification.

Catalog No. 50-002-59287 Supplier Medchemexpress LLC Supplier No. HY13443310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GDP-L-fucose disodium is a nucleotide sugar that serves as the fucose donor in the biosynthesis of fucose-containing oligosaccharides and is used as a substrate in enzymatic and biochemical assays. The material is supplied as a high-purity solid suitable for analytical standards and enzyme reaction studies.

  • High purity (99.8%) for reliable experimental results.
  • Chemical formula C16H25N5O15P2 for precise molecular identification.
  • Supplied as a disodium salt for good water solubility and ease of use.
  • Suitable as a substrate for fucosyltransferase and glycosylation assays.
  • Available in small-scale quantities appropriate for research applications.
  • CAS number 15839-70-0 provided for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.