Learn More
Cayman Chemical GDP-L-Fucose Synthasehumn100ug
SDP
Catalog No. 502936928
Encompass_Preferred
Source: Recombinant N-terminal histidine-tagged GDP-L-fucose synthase purified from E. coli • Amino acids: 2-321 • MW: 37.98 kDa
Catalog No. 50-293-6928 Supplier Cayman Chemical Supplier No. 26695100 UG
Source: Recombinant N-terminal histidine-tagged GDP-L-fucose synthase purified from E. coli • Amino acids: 2-321 • MW: 37.98 kDa