Learn More
Medchemexpress LLC Gdp366 | 501698-03-9 | 375.45 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYU0017710MG
Encompass_Preferred
GDP366 is a dual inhibitor of survivin and Op18 that induces cell growth inhibition, cellular senescence, and mitotic catastrophe in human cancer cells.
  • Potently and selectively inhibits survivin and Op18 expression.
  • Decreases survivin and Op18 mRNA and protein levels.
  • Significantly inhibits tumor cell growth in vitro and in vivo.
  • Induces polyploidy and increases chromosomal instability in cancer cells.
  • Promotes cellular senescence by inhibiting telomerase activity.
Catalog No. 50-004-49661