Learn More
Medchemexpress LLC HY-U00177 5mg Medchemexpress, GDP366 CAS:501698-03-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001775MG
Encompass_Preferred
Medchemexpress, HY-U00177 5mg GDP366 CAS:501698-03-9 GDP366, a dual inhibitor of survivin and Op18 , induces cell growth inhibition, cellular senescence and mitotic catastrophe in human cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4432