Learn More
Perkin Elmer US LLC Germanium Crystal with Holder - Spare
Catalog No. 509043408
Encompass_Preferred
This crystal and holder are compatible with the ATR Objective for the i-Series and AutoIMAGE System Microscopes.
Catalog No. 50-904-3408 Supplier Perkin Elmer US LLC Supplier No. L1860268
This crystal and holder are compatible with the ATR Objective for the i-Series and AutoIMAGE System Microscopes.