Learn More
Bulbman Inc GE F15T8BL BLACKLIGHT 15W 18IN
SDP
Catalog No. 501253577
Encompass_Preferred
GE F15T8/BL: T8 "Blacklight" fluorescent lamp. 15 Watts and 18" for use in smaller fixtures. * Produces UV-A Light often used for Nail Polish Curing, Chemical Processing, and Insect Trap Applications
Catalog No. 50-125-3577 Supplier Bulbman Inc Supplier No. 112383
GE F15T8/BL: T8 "Blacklight" fluorescent lamp. 15 Watts and 18" for use in smaller fixtures. * Produces UV-A Light often used for Nail Polish Curing, Chemical Processing, and Insect Trap Applications