Learn More
Enzo Life Sciences PGE2 FPIA kit (1x100tests)
SDP
Supplier:  Enzo Life Sciences ADI920001
Encompass_Preferred
The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8. Alternative Name: Prostaglandin E2. Sensitivity: 684 pg/ml (range 1,562 - 100,000 pg/ml). Assay Time: 30 minutes. Applications: Fluorescence microscopy. Species Reactivity: Species independent. Contents: Conjugate concentrate, Antibody, Assay buffer concentrate, Standard
Catalog No. 50-658-663