Learn More
Enzo Life Sciences GEA 5024 (10mg). CAS: 144575-27-9
SDP
Catalog No. 502009129
Encompass_Preferred
Water soluble and stable nitric oxide donor. Alternative name: 5-Amino-3-(3-chloro-2-methylphenyl)-1,2,3,4-oxatriazolium chloride. Purity: ≥97% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9129 Supplier Enzo Life Sciences Supplier No. ALX430005M010
Water soluble and stable nitric oxide donor. Alternative name: 5-Amino-3-(3-chloro-2-methylphenyl)-1,2,3,4-oxatriazolium chloride. Purity: ≥97% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in water. Long Term Storage: +4°C.