Learn More
Enzo Life Sciences GEA 5583 (10mg)
SDP
Catalog No. 502009130
Encompass_Preferred
Stable nitric oxide releasing compound that is orally absorbed in rats. Alternative name: 1,2,3,4-Oxatriazolium, 3-(3-chloro-2-methylphenyl)-5-(((cyanomethylamino)carbonyl)amino)-, hydroxide inner salt. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-9130 Supplier Enzo Life Sciences Supplier No. ALX430006M010
Stable nitric oxide releasing compound that is orally absorbed in rats. Alternative name: 1,2,3,4-Oxatriazolium, 3-(3-chloro-2-methylphenyl)-5-(((cyanomethylamino)carbonyl)amino)-, hydroxide inner salt. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.