Learn More
Medchemexpress LLC GeA-69 | 2143475-98-1 | 99.9% | 300.35 | 10 MG
SDP
Catalog No. 5000346597
Encompass_Preferred
GeA-69 is a selective, allosteric inhibitor of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14) targeting macrodomain 2 (MD2). It is involved in DNA damage repair mechanisms and prevents the recruitment of PARP14 MD2 to sites of laser-induced DNA damage. This product is for research use only.
  • Selective, allosteric inhibitor of PARP14 targeting MD2.
  • Involved in DNA damage repair mechanisms.
  • Prevents recruitment of PARP14 MD2 to sites of laser-induced DNA damage.
  • Highly cell permeable.
  • Exhibits moderate cytotoxicity against normal cells.
Catalog No. 50-003-46597 Supplier Medchemexpress LLC Supplier No. HY10870810MM/1ML
GeA-69 is a selective, allosteric inhibitor of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14) targeting macrodomain 2 (MD2). It is involved in DNA damage repair mechanisms and prevents the recruitment of PARP14 MD2 to sites of laser-induced DNA damage. This product is for research use only.
  • Selective, allosteric inhibitor of PARP14 targeting MD2.
  • Involved in DNA damage repair mechanisms.
  • Prevents recruitment of PARP14 MD2 to sites of laser-induced DNA damage.
  • Highly cell permeable.
  • Exhibits moderate cytotoxicity against normal cells.