Learn More
Arrow Engineering Gear DrivenOverhead Stirrer, Electric Model 850, 120V 60HZ
Catalog No. NC3796500
Encompass_Preferred
Arrow Mixing Products Model 850 stands out for its exceptional performance and reliability. It’s engineered for medium to heavy-duty mixing tasks, making it ideal for a variety of industrial applications. With its high torque and speed range of 20-850 RPM's, this electric overhead stirrer can handle large batches with ease. The Model 850 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.
Catalog No. NC3796500 Supplier Arrow Engineering Supplier No. MODEL 850
Arrow Mixing Products Model 850 stands out for its exceptional performance and reliability. It’s engineered for medium to heavy-duty mixing tasks, making it ideal for a variety of industrial applications. With its high torque and speed range of 20-850 RPM's, this electric overhead stirrer can handle large batches with ease. The Model 850 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.