Learn More
AVANTOR MASTERFLEX (VWR LLC) GEARPMP SYSTM CVTY 0.91ML/REV
Catalog No. 502742111
Encompass
Masterflex Ismatec Reglo Gear Pump System with MasterflexLive, 4000 rpm Drive, A-Mount Cavity-Style Pump Head, 0.91 mL/rev; 115/230 VAC, Avantor
Catalog No. 50-274-2111 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7801886
Masterflex Ismatec Reglo Gear Pump System with MasterflexLive, 4000 rpm Drive, A-Mount Cavity-Style Pump Head, 0.91 mL/rev; 115/230 VAC, Avantor