Learn More
MSC GEARWRENCH Impact Socket, 1/2 in, 1-5/16 in, 6 Point, Deep
Catalog No. 5000508724
Encompass
This impact socket is designed for heavy-duty applications, featuring a 1/2 inch drive and a 1-5/16 inch 6-point deep configuration. Constructed from durable Chromium-Molybdenum Steel with a black oxide finish, it offers reliable performance for fastening tasks.
  • High-visibility laser-etched markings provide quick size identification.
  • Off-corner loading design reduces fastener rounding.
  • Secondary hard stamped size markings.
Catalog No. 50-005-08724 Supplier MSC Supplier No. 38057378
This impact socket is designed for heavy-duty applications, featuring a 1/2 inch drive and a 1-5/16 inch 6-point deep configuration. Constructed from durable Chromium-Molybdenum Steel with a black oxide finish, it offers reliable performance for fastening tasks.
  • High-visibility laser-etched markings provide quick size identification.
  • Off-corner loading design reduces fastener rounding.
  • Secondary hard stamped size markings.