Learn More
MSC GEARWRENCH Torx Bit Socket Set, 10 Piece, 1/4 in & 3/8 in Drive, T10-T55
Catalog No. 5000507767
Encompass
This 10 piece 1/4 in and 3/8 in drive Torx bit socket set comes on a socket rail for storage and organization. The bits are made from S2 steel for strength and are heat-treated for durability. Each socket has hard-stamped sizes for easy identification, and features a bright, full polish chrome finish that resists corrosion. This set meets or exceeds ASME specifications.
  • Bits made of S2 steel for strength and durability.
  • Heat treated for maximum durability.
  • Hard stamped size markings for easy identification.
  • Bright, full polish chrome finish resists corrosion.
  • Includes socket rail for convenient storage and organization.
  • Meets or exceeds ASME specifications.
Catalog No. 50-005-07767 Supplier MSC Supplier No. 86567815
This 10 piece 1/4 in and 3/8 in drive Torx bit socket set comes on a socket rail for storage and organization. The bits are made from S2 steel for strength and are heat-treated for durability. Each socket has hard-stamped sizes for easy identification, and features a bright, full polish chrome finish that resists corrosion. This set meets or exceeds ASME specifications.
  • Bits made of S2 steel for strength and durability.
  • Heat treated for maximum durability.
  • Hard stamped size markings for easy identification.
  • Bright, full polish chrome finish resists corrosion.
  • Includes socket rail for convenient storage and organization.
  • Meets or exceeds ASME specifications.