Learn More
TARGETMOL CHEMICALS INC GEDUNIN 5MG
SDP
Catalog No. 502580991
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gedunin is an important limonoid present in several genera of the Meliaceae family mainly in seeds. Gedunin is an Hsp90 inhibitor and inhibits ovarian cancer cell proliferation. purity: 99%
Catalog No. 50-258-0991 Supplier TARGETMOL CHEMICALS INC Supplier No. T218835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gedunin is an important limonoid present in several genera of the Meliaceae family mainly in seeds. Gedunin is an Hsp90 inhibitor and inhibits ovarian cancer cell proliferation. purity: 99%