Learn More
Medchemexpress LLC Gefapixant citrate | 2310299-91-1 | 99.2% | 545.5 g/mol | C20H27N5O11S | 1 ML
SDP
Catalog No. 5000249109
Encompass_Preferred
Gefapixant citrate is a P2X3 receptor antagonist provided as a concentrated solution for laboratory research. The reagent is supplied at 10 mM in DMSO (1 mL) and is intended for in vitro pharmacology and biochemical studies, with analytical documentation available from the manufacturer.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.2%) suitable for research applications.
  • Supports in vitro P2X3 receptor pharmacology and biochemical assays.
  • Comes with data sheet, certificate of analysis, and safety data sheet.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-49109 Supplier Medchemexpress LLC Supplier No. HY101588A10MM 1ML
Gefapixant citrate is a P2X3 receptor antagonist provided as a concentrated solution for laboratory research. The reagent is supplied at 10 mM in DMSO (1 mL) and is intended for in vitro pharmacology and biochemical studies, with analytical documentation available from the manufacturer.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.2%) suitable for research applications.
  • Supports in vitro P2X3 receptor pharmacology and biochemical assays.
  • Comes with data sheet, certificate of analysis, and safety data sheet.
  • For research use only; not for human or diagnostic use.