Learn More
Medchemexpress LLC Gefapixant citrate | 2310299-91-1 | 99.2% | 545.52 g/mol | C20H27N5O11S | 5 MG
SDP
Catalog No. 5000245671
Encompass_Preferred
Gefapixant citrate is the citrate salt of gefapixant, an orally active P2X3 receptor antagonist used in research on chronic cough and pain-related indications. Supplied as a research-grade solid with high purity, it is suitable for in vitro and in vivo studies and is accompanied by technical documentation for analytical use.
  • Potent P2X3 receptor antagonist with low-nanomolar activity.
  • High purity suitable for research applications.
  • Available in small solid quantities for dose-ranging studies.
  • Includes datasheet, certificate of analysis, and safety data sheet documentation.
  • Soluble stock solutions offered for convenient preparation.
Catalog No. 50-002-45671 Supplier Medchemexpress LLC Supplier No. HY101588A5MG
Gefapixant citrate is the citrate salt of gefapixant, an orally active P2X3 receptor antagonist used in research on chronic cough and pain-related indications. Supplied as a research-grade solid with high purity, it is suitable for in vitro and in vivo studies and is accompanied by technical documentation for analytical use.
  • Potent P2X3 receptor antagonist with low-nanomolar activity.
  • High purity suitable for research applications.
  • Available in small solid quantities for dose-ranging studies.
  • Includes datasheet, certificate of analysis, and safety data sheet documentation.
  • Soluble stock solutions offered for convenient preparation.