Learn More
Medchemexpress LLC HY-50895 1g , Gefitinib CAS:184475-35-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY508951G
Encompass_Preferred
HY-50895 1g Gefitinib CAS: 184475-35-2 Gefitinib (ZD1839) is a potent, selective and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. Gefitinib selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and that blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib also induces autophagy and cell apoptosis, which can be used for cancer related research, such as Lung cancer and breast cancer [5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9084