Learn More
AdipoGen Gefitinib,Chemical. CAS: 184475-35-2
SDP
Catalog No. 501016269
Encompass_Preferred
Gefitinib, AdipoGen Life Sciences. Chemical. CAS: 184475-35-2. Formula: C22H24ClFN4O3. MW: 446.9. Gefitinib is a selective, orally available epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor. In NR6wtEGFR and NR6W cells, it inhibits Tyr1173, Tyr992, Tyr1173 and Tyr992 with IC(50) values of 37nM, 37nM, 26nM and 57nM, respectively.
Catalog No. 50-101-6269 Supplier AdipoGen Supplier No. SYN1042M001
Gefitinib, AdipoGen Life Sciences. Chemical. CAS: 184475-35-2. Formula: C22H24ClFN4O3. MW: 446.9. Gefitinib is a selective, orally available epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor. In NR6wtEGFR and NR6W cells, it inhibits Tyr1173, Tyr992, Tyr1173 and Tyr992 with IC(50) values of 37nM, 37nM, 26nM and 57nM, respectively.