Learn More
Medchemexpress LLC HY-50895 5g , Gefitinib ZD1839 CAS:184475-35-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY508955G
Encompass_Preferred
Medchemexpress, HY-50895 5g Gefitinib ZD1839 CAS:184475-35-2 Formula:C22H24ClFN4O3 IC50: 37 nM (Tyr1173 site, in NR6wtEGFR cells), 37 nM (Tyr992 site, in NR6wtEGFR cells) Purity:98% Gefitinib is an inhibitor that specifically binds and inhibits the EGFR tyrosine kinase , with the IC 50 value of 2-37 nM in NR6wtEGFR cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2170