Learn More
Medchemexpress LLC HY-50895 500mg Medchemexpress, Gefitinib CAS:184475-35-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50895500MG
Encompass_Preferred
Medchemexpress, HY-50895 500mg Gefitinib CAS:184475-35-2 Gefitinib (ZD1839) is a potent, selective and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. Gefitinib selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and that blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib also induces autophagy. Gefitinib has antitumour activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9348