Learn More
AdipoGen Gefitinib,Chemical. CAS: 184475-35-2
SDP
Supplier:  AdipoGen SYN1042M050
Encompass_Preferred
Gefitinib, AdipoGen Life Sciences. Chemical. CAS: 184475-35-2. Formula: C22H24ClFN4O3. MW: 446.9. Gefitinib is a selective, orally available epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor. In NR6wtEGFR and NR6W cells, it inhibits Tyr1173, Tyr992, Tyr1173 and Tyr992 with IC(50) values of 37nM, 37nM, 26nM and 57nM, respectively.
Catalog No. 50-101-6272